Learn More
Essendant CLEANERINDUSTRIAL12/CS
Supplier:  Essendant AMR1002262
Encompass_Preferred
Cleaning is quick and simple without the danger of injury to working parts with energized electrical cleaner Fast dry non-flammable chlorinated formula rapidly removes dirt grease oil and grime from metallic and nonmetallic surfaces Ideal for use on electric motors (even while running) electronic equipment ignition systems typewriters computers etc Safe on most plastics Application- Electronics/Antistatic Cleaner Applicable Material- Ceramics Fiberglass Glass Laminates Metal Plastic Dirt Types- Debris Dust Grease Fingerprints Soil Smudges Scent- Unscented
Catalog No. 50-000-22746