Learn More
Essendant CLEANEROVEN
Supplier:  Essendant MLB25000041
Encompass_Preferred
Great for decarbonizing cooking utensils ready-to-use no mixing of harsh or dangerous chemicals Thick rich foam stays where you spray it allowing this outstanding cleaning chemical the time needed to break down even the worst soils Convenient and easy to use just pop a foaming trigger sprayer into a quart and you are ready to go Application- Kitchens Applicable Material- Ovens Grills Chemical Compound- Potassium Hydroxide Dirt Types- Carbonized Soils
Catalog No. 50-000-33849