Learn More
Essendant CLEANEROVN/GRL32OZ12/C
Supplier:  Essendant DVO948049
Encompass_Preferred
High-powered product removes grease and heavily carbonized soils Oil- and grime- cutting additives minimize the need for labor-intensive scrubbing May be diluted for use on fryers Application- Degreaser/Cleaner Applicable Material- Ceramics Enamel Glass Metal Plastic Dirt Types- Carbon Food Debris Grease Grime Oil-Based Soil Organic Matter Smoke Film Scent- Unscented
Catalog No. 50-000-21769