Learn More
Essendant CLEANERPOLISH32OZ
Supplier:  Essendant EOPPL93306EA
Encompass_Preferred
The ECOS PRO reliable and effective water-based formula cleans polishes and protects stainless steel surfaces The cleaner will not leave streaks hazing or oily residues behind Protects against water spots and fingerprints and formulated to perform without harsh fumes Excellent on other metal surfaces such as aluminum brass chrome copper zinc silver and mild steel The pH-balanced formula is made using sustainable plant-powered ingredients is free of dyes and is gray water and septic safe Not tested on animals Manufactured in a facility that is LEED Zero Carbon LEED Zero Waste and LEED Zero Energy Certified Application- Metal Cleaner/Polish Applicable Material- Aluminum Brass Chrome Copper Silver Stainless Steel Zinc Chemical Compound- Water Methyl Soyate Laureth 9 Laureth 3 Phenoxyethanol Methylisothiazolinone Dirt Types- Finger Prints Water Spots
Catalog No. 50-000-35478