Learn More
Hach Company Cleaning Brush for Large Capacity Sludge Judge
Supplier:  Hach Company 2506103
Encompass
Sludge Judge accessories help maintain a long, trouble-free service life. Cleaning brush not recommended for cleaning oil and grease. Use Grease Trap Cleaning Rod in conjunction with Cleaning Strips and a mild detergent to effectively remove oil & grease from plastic tubing.
Catalog No. 50-199-6108