Learn More
Sklar Instruments Cannula Instrument Cleaning Brush, 3mm x 12 in., Pack of 3.
SDP
Supplier:  Sklar Instruments 101660
Encompass_Preferred
Cannula Instrument Cleaning Brush, 3mm x 12 in., Pack of 3. Cannula Instrument Cleaning Brushes are used in the multiple-step process of sterilizing a reusable cannula. Following soaking in enzymatic solution, the cannula should be cleaned manually with a brush and detergent or ultrasonically. This product is 3mm x 12 inches.
Catalog No. 50-110-9725