Learn More
Brady™ CleanLift™ Series Thermal Transfer Printable Labels, B-449
Ideal for a variety of applications. Brady™ CleanLift™ Series Thermal Transfer Printable Labels are made of removable polypropylene
Supplier:  Brady™ THT174493
 View more versions of this product
Catalog No. 19-109-248
Add to Cart
Description
Description
  • Available in various sizes
Specifications
Specifications
Recommended ribbon series: R6200
None
Matte
Polypropylene (B-449)
Thermal Transfer
Non-sterile
BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
-70°C
3000 Label(s) / Roll
Rectangle
50.8 x 25.4 mm (W x H)
-70°C to +90°C
0.112 mm
25.40 x 50.80 mm
2 in.
50.80 mm
25.4 mm
Acrylic, Removable
White
Roll
Single-side Printable
Thermal Transfer Printer Label
Barcode labeling, Component and equipment identification, General and industrial identification
1
THT-17-449-3
3000/Pk.
50.80 mm (W) x 25.40 mm (H)
-94°F to 194°F
0.0044 in.
28.58 mm
1 in.
50.8 mm
1 in.
THT-17-449-3
Product Suggestions
Product Suggestions