Learn More
Med Vet International SafeWash™ Irrigation Solution, Sodium Chloride 0.9%, 7.1-ounce Spray Can, 00245, Each
Supplier:  Med Vet International 670708EA
Encompass_Preferred
The sodium chloride wound wash comes in a can with point and spray technology and can be used upright or upside down It provides irrigation pressure to effectively remove debris and bacteria from wounds The saline wound wash is free of drugs and preservatives It will not sting or burn for greater user comfort Use it to cleanse minor wounds, puncture wounds, deep cuts, scrapes and for first aid and incontinence care Spray can contains 209.79 grams of wound cleanser /ul>
Catalog No. 50-304-3723