Learn More
GA International Clear Double-Sided Tape for Frozen Surfaces with GatorCUT™ Technology (Patented) 2" x 100' / 50.8mm x 30m Transparent 3" / 76.2mm (plastic) From -80°C to +90°C (-112°F to +194°F)
Supplier:  GA International FST50CP3100
Encompass_Preferred
Our clear double-sided tape is a specialized tape designed for use in ultra-low temperature environments. Its specially formulated permanent adhesive retains its bonding properties even at extremely low temperatures, such as those found in laboratory freezers (as low as -80°C) and during transport on dry ice. The tape is designed to bond securely to plastic bags, even in harsh, deep-freeze conditions.FrozenSTUCK™ tape can be easily carried and handled due to our unique GatorCUT technology, which allows it to be cut easily without the need for scissors, dispensers, or other cutting instruments.This deep-freeze double-sided tape for plastic bags is ideal for use in biomedical research and laboratory work involving storing and transporting biological samples at extremely low temperatures.FrozenSTUCK tape provides a reliable and effective solution for any applications requiring secure bonding in harsh deep-freeze conditions.
Catalog No. NC3823043