Learn More
Carbosynth LLC. CLEAR-OX 5g
SDP
Supplier:  Carbosynth LLC. RCO1260PI5G
Encompass_Preferred
Application: Polymer-Supported Oxidant for Disulfide Bond Formation CLEARâ„¢ Polymer-Supported Oxidant for the Facile Formation of Disulfide-Bridged Peptides, Storage Temperature: 2-10°C
Catalog No. 50-168-6857