Learn More
Carbosynth LLC. CLEAR-OX, 1g
SDP
Supplier:  Carbosynth LLC. RCO1260PI1G
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Application: Polymer-Supported Oxidant for Disulfide Bond Formation CLEAR Polymer-Supported Oxidant for the Facile Formation of Disulfide-Bridged Peptides, Storage Temperature: 2-10C
Catalog No. 50-168-6856