Learn More
Molecular Dimensions CLEAR STRATEGY SCREEN II HT-96
SDP
Supplier:  Molecular Dimensions MD132
Encompass
Protein Crystallography. Clear Strategy Screen II HT-96 (96 x 1.1mL): The companion screen to Clear Strategy Screen I. Allows users to easily interpret results & plan optimization. In the HT-96 format, the Clear Strategy II reagents are premixed at 4 pH values (pH's 5.5, 6.5, 7.5 and 8.5).
Catalog No. NC0090594