Learn More
Ethos Biosciences Clear-Sub, Xylene Substitute, 1gal, Ethos
Supplier:  Ethos Biosciences 3344G
Encompass_Preferred
Xylene Substitute is a clear, high-performance clearing agent designed for pathology. This naphthenic solvent is formulated as a safer alternative to traditional xylene, it is ideal for tissue processing, slide clearing, and coverslipping procedures. The solution provides effective paraffin removal and excellent optical clarity for reliable microscopic evaluation. With reduced odor and improved handling characteristics, it supports a more comfortable laboratory environment while maintaining consistent performance.
Catalog No. 50-005-74169