Learn More
Ethos Biosciences Xylene, 1gal, Ethos
Supplier:  Ethos Biosciences 3346G
Encompass_Preferred
Xylene is an aromatic solvent suitable for pathology and general laboratory applications. Commonly used as a clearing agent in tissue processing, it is ideal for paraffin removal, slide clearing, and coverslipping procedures. The clear, colorless solution provides excellent optical clarity for reliable microscopic evaluation. Each batch is tested for moisture to help ensure consistent performance and dependable results. Xylene is also widely used for solvent extraction, cleaning, and other routine laboratory tasks. Supplied in chemically compatible containers, it is appropriate for clinical, research, pharmaceutical, and industrial laboratories.
Catalog No. 50-005-74314