Learn More
Alkali Scientific ClearSeal™ 2.0mL Generic Screw Cap Micro Tubes, without Graduations, RNase, DNase & Pyrogen Free, without Cap, 1000/Cs
SDP
Supplier:  Alkali Scientific MT3472
Encompass_Preferred
ClearSeal™ 2.0mL screw cap microtubes are engineered for performance and reliability in high demand laboratory environments. Made from ultra clear polypropylene, they provide exceptional visibility of samples while ensuring maximum safety and sample integrity. Each tube comes pre assembled with a silicone O ring screw cap, delivering a leak proof seal capable of withstanding temperatures from 90°C to +121°C. Ideal for a wide range of applications, including cryogenic storage and high speed centrifugation. Key Features: Ultra Clear Polypropylene: Ensures optimal sample visibility for accurate inspection. Leak Proof Seal, Assembled caps with silicone O rings provide a secure seal across extreme temperatures ( 90°C to +121°C). Certified Sterile or Free from RNase, DNase, and pyrogens for contamination free performance. High Speed Centrifugation Rated for centrifugation up to 22,000×g. Flexible Design Options, Available with white, printed graduations or as non gr
Catalog No. 50-006-62247