Learn More
Cleatech ACRYLIC GLOVE BOX W/ AIR LOCK
SDP
Supplier:  Cleatech 22002AGGT
Encompass_Preferred
Two-Port Portable Mini Glovebox, non-vacuum, Clear Acrylic, 27"Wx 18"Dx 18"H, one swing up access door with opening of 8"x18", 8" Diameter glove ports with stainless steel clamp. Including Air Lock (transfer Chamber 11.5"Wx 10.5"Dx10.5"H) ,Including Gas In Port and Pressure relief Valve , Including 15 mil Chemical Resistance Gloves
Catalog No. 50-152-7898