Learn More
ABclonal Technology Cleaved Caspase-7 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2315420UL
Encompass_Preferred
This gene encodes a member of the cysteine-aspartic acid protease (caspase) family Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits large and small that dimerize to form the active enzyme The precursor of the encoded protein is cleaved by caspase 3 and 10 is activated upon cell death stimuli and induces apoptosis Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene
Catalog No. 50-000-06375