Learn More
Medchemexpress LLC Clebopride malate | 57645-91-7 | 99.4% | 1 ML
Supplier: Medchemexpress LLC HYB1613A10MM/1ML
Clebopride malate is an orally active dopamine receptor antagonist. It acts on dopamine D2 receptors and exhibits antiemetic and prokinetic effects. This compound is suitable for research into functional gastrointestinal disorders.
- Orally active dopamine receptor antagonist
- Acts on dopamine D2 receptors
- Possesses antiemetic effects
- Exhibits prokinetic effects
- Selectively binds to D2 dopamine receptors and α2 adrenergic receptors