Learn More
Medchemexpress LLC Clec11a/SCGF-alpha protein, rat (HEK293, His) | ≥90.0% | 100UG

Supplier: Medchemexpress LLC HYP703697100UG
Recombinant, glycosylated rat CLEC11A (SCGF-alpha) produced in HEK293 cells with a C-terminal His tag, supplied as a sterile PBS solution for research applications.
- Glycosylated protein with apparent molecular weight approximately 45-60 kDa.
- Purity ≥90% by reducing SDS-PAGE.
- Supplied in PBS, pH 7.4, 0.22 μm filtered solution at 0.57 mg/mL.
- Contains a C-terminal His tag for detection and purification.
- Recommended storage at -80°C; avoid repeated freeze-thaw cycles.
- Suitable for in vitro biochemical and cell-based assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.