Learn More
Aviva Systems Biology Corp CLEC6A Antibody - N-terminal region (ARP44533_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44533_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLEC6A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CLEC6A. Alternate names: CLECSF10, dectin-2, CLEC4NApplicationWB. Species reactivity: Cow, Human, Mouse, Rat
Catalog No. 50-014-53988