Learn More
Medchemexpress LLC Clematichinenoside AR | 761425-93-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN42325MG
Encompass_Preferred
Clematichinenoside AR is a major active ingredient extracted from the traditional Chinese herb *Clematis chinensis*. It exhibits potent pharmacological effects on various diseases, including atherosclerosis, by activating autophagy to alleviate foam cell formation and inflammatory responses in Ox-LDL-induced RAW264.7 cells.
  • Alleviates foam cell formation
  • Reduces inflammatory response
  • Activates autophagy
  • For research use only
Catalog No. 50-004-20812