Learn More
Apexbio Technology LLC Clemizole 442-52-4 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A33155.1
Encompass_Preferred
Clemizole (CAS 442-52-4) is a small-molecule antagonist targeting the histamine H1 receptor and the hepatitis C virus (HCV) NS4B protein It is designed to block the H1 receptor and disrupt NS4B-associated RNA-binding thereby inhibiting viral replication Clemizole exerts its biological activity primarily through antagonism of the H1 receptor and interference with NS4B-mediated replication processes In in vitro assays clemizole demonstrates inhibitory activity with an IC50 value of approximately 8 mM against HCV genotype 2a replication Based on these pharmacological properties clemizole holds research potential in anti-HCV therapy and related antiviral applications
Catalog No. 50-000-95290