Learn More
Medchemexpress LLC Clenbuterol hydrochloride | 21898-19-1 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB16145MG
Encompass_Preferred
Clenbuterol (NAB-365) hydrochloride is a selective β2-adrenergic agonist that enhances skeletal muscle strength and hypertrophy. It also induces growth factor mRNA, activates astrocytes, and protects rat brain tissue against ischemic damage.
  • Selective β2-adrenergic agonist
  • Enhances skeletal muscle strength and hypertrophy
  • Induces growth factor mRNA
  • Activates astrocytes
  • Protects rat brain tissue against ischemic damage
Catalog No. 50-004-13124