Learn More
Medchemexpress LLC Cleomiscosin A | 76948-72-6 | 10 MM * 1 ML

Supplier: Medchemexpress LLC HYN359510MM/1ML
Cleomiscosin A is a coumarino-lignoid derived from the branch of Macaranga adenantha. It is active against TNF-alpha secretion of mouse peritoneal macrophages.
- Purity of 99.15%
- Appears as a solid, white to off-white
- Molecular weight: 386.35
- Formula: C20H18O8
- For research use only
- Inhibits LPS-induced NO production in mouse BV2 cells
- Cytotoxic against human KB cells (ED50 > 10 μg/mL) and mouse P388 cells (ED50 0.4 μg/mL)
- Soluble in DMSO (125 mg/mL) and corn oil (≥ 2.08 mg/mL with 10% DMSO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.