Learn More
TARGETMOL CHEMICALS INC CLETOQUINE HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T10835L15MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cletoquine hydrochloride is a major active metabolite of Hydroxychloroquine. Cletoquine hydrochloride is a Chloroquine derivative and exhibits the ability to against the chikungunya virus (CHIKV). Cletoquine hydrochloride exhibits antimalarial effects. purity: 98%
Catalog No. 50-258-1649