Learn More
Selleck Chemical LLC Clevudine S3001-50mg
SDP
Supplier:  Selleck Chemical LLC S300150MG
Encompass_Preferred
Clevudine(Levovir L-FMAU) is an antiviral drug for the treatment of hepatitis B
Catalog No. 50-311-2434