Learn More
Medchemexpress LLC Clevudine (L-FMAU) | 163252-36-6 | 99.8% | 260.22 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1385950MG
Encompass_Preferred
Clevudine (L-FMAU) is a nucleoside analog of the unnatural L-configuration with potent anti-HBV activity, long half-life, and low toxicity. It acts as a non-competitive inhibitor that binds to the polymerase rather than being incorporated into the viral DNA. Clevudine is also active against cowpox virus respiratory infection in mice.
  • Potent anti-HBV activity
  • Long half-life
  • Low toxicity
  • Acts as a non-competitive inhibitor
  • Active against cowpox virus respiratory infection in mice
Catalog No. 50-003-49840