Learn More
ABclonal Technology CLIC1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A136841000UL
Encompass_Preferred
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 1 is a member of the p64 family, the protein localizes principally to the cell nucleus and exhibits both nuclear and plasma membrane chloride ion channel activity.
Catalog No. 50-000-97663