Learn More
Medchemexpress LLC Clinafloxacin (hydrochloride) | 105956-99-8
SDP
Supplier:  Medchemexpress LLC 25MGHYB0536
Encompass_Preferred
Clinafloxacin hydrochloride is a potent and broad-spectrum fluoroquinolone antibiotic. It demonstrates inhibitory activity against gram-positive and gram-negative bacteria, as well as anaerobic pathogens, in vitro.
  • Potent and broad-spectrum fluoroquinolone antibiotic
  • Inhibitory activity against gram-positive bacteria
  • Inhibitory activity against gram-negative bacteria
  • Inhibitory activity against anaerobic pathogens
  • Targets DNA gyrase and topoisomerase IV of *S. aureus*
  • Active in antibiotic and infection research
Catalog No. 50-004-50520