Learn More
Medchemexpress LLC Clinafloxacin hydrochloride | 105956-99-8 | 99.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0536A100MG
Encompass_Preferred
Clinafloxacin hydrochloride is a potent and broad-spectrum fluoroquinolone antibiotic. It exhibits inhibitory activity against gram-positive, gram-negative bacteria, and anaerobic pathogens in vitro. It targets DNA gyrase and topoisomerase IV of S. aureus with IC50 values of 0.92 μg/ml and 1.62 μg/ml, respectively.
  • Quinolone IC50 and target
  • Active against S. pneumonia with an MIC of 1 μg/ml for parC-gyrA mutants
  • Exhibits antibacterial activities against target-altered mutant strains of S. aureus
  • Effective in treating penicillin-resistant, ciprofloxacin-susceptible pneumococcal meningitis in a rabbit model
Catalog No. 50-004-08880