Learn More
Medchemexpress LLC Clindamycin | 18323-44-9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1455100MG
Encompass_Preferred
Clindamycin is an orally active, broad-spectrum bacteriostatic lincosamide antibiotic. It inhibits bacterial protein synthesis and can suppress the expression of virulence factors in *Staphylococcus aureus* at sub-inhibitory concentrations (sub-MICs). Resistance to Clindamycin occurs due to enzymatic methylation of the antibiotic binding site in the 50S ribosomal subunit (23S rRNA). Clindamycin also reduces the production of Panton-Valentine leucocidin (PVL), toxic-shock-staphylococcal toxin (TSST-1), or alpha-haemolysin (Hla), and can be used in malaria research.
  • Orally active
  • Broad-spectrum bacteriostatic lincosamide antibiotic
  • Inhibits bacterial protein synthesis
  • Suppresses virulence factor expression in *Staphylococcus aureus*
  • Decreases production of PVL, TSST-1, or Hla
  • Can be used for malaria research
Catalog No. 50-004-12699