Learn More
Medchemexpress LLC Clindamycin phosphate | 24729-96-2 | MFCD11982855 | 99.5% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB106410MM/1ML
Encompass_Preferred
Clindamycin phosphate (Clindamycin 2-phosphate) is a broad-spectrum bacteriostatic lincosamide antibiotic. It is a proagent of Clindamycin that lacks in vitro antimicrobial activity but converts rapidly in vivo to the active parent agent, Clindamycin, through phosphatase ester hydrolysis. It is used in research for conditions like acne and bacterial vaginosis, and it exhibits no cytotoxicity. When combined with platelet-rich fibrin (PRF), PRF-Clindamycin phosphate enhances antimicrobial properties.
  • Broad-spectrum bacteriostatic lincosamide antibiotic.
  • Proagent of Clindamycin, converting to the active form in vivo.
  • Can be used for researching acne and bacterial vaginosis.
  • No cytotoxicity observed.
  • Enhances antimicrobial properties when combined with platelet-rich fibrin.
Catalog No. 50-004-14036