Learn More
Medchemexpress LLC Clindma | 908860-82-2 | MFCD00382831 | 99.9% | 808.35 | C54H97NO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1127635MG
Encompass_Preferred
CLinDMA is a cationic lipid used in research to formulate lipid nanoparticles for nucleic acid delivery. It is supplied for research use only and has been reported to induce inflammatory responses; users should consult the batch COA and SDS for exact purity and handling instructions.
  • Cationic lipid for lipid nanoparticle formulation.
  • Applicable to siRNA and other nucleic acid delivery research.
  • Reported high purity but supplier-dependent; verify with batch COA.
  • Soluble in DMSO and offered as solid or ready-to-use solution (e.g., 10 mM in DMSO).
  • Reported appearance: light yellow to yellow viscous liquid.
Catalog No. 50-002-62971