Learn More
Medchemexpress LLC Clioquinol | 130-26-7 | C9H5CLINO | 5 G
SDP
Supplier:  Medchemexpress LLC HY146035G
Encompass_Preferred
Clioquinol (Iodochlorhydroxyquin) is a topical antifungal agent with anticancer activity. It acts as an oral antimicrobial for the research of diarrhea and skin infections. Clioquinol is a chelator that binds to Zn²⁺ and Cu²⁺ with a greater affinity than to Ca²⁺ and Mg²⁺, and it can freely cross the blood-brain barrier for the study of Alzheimer's disease.
  • Topical antifungal agent
  • Exhibits anticancer activity
  • Functions as an oral antimicrobial for diarrhea and skin infections research
  • Acts as a chelator binding to Zn²⁺ and Cu²⁺
  • Freely crosses the blood-brain barrier
  • Used for the study of Alzheimer's disease
  • Relevant for research in fungal, autophagy, mitophagy, antibiotic, and parasite studies
  • Applicable in infection, neurological disease, inflammation/immunology, and cancer research
Catalog No. 50-003-56270