Learn More
Enzo Life Sciences CLIP monoclonal antibody (CerCLIP.1) (200µg)
SDP
Supplier:  Enzo Life Sciences ADICSA600F
Encompass_Preferred
Clone: CerCLIP.1. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of human CLIP. Source: Purified from mouse ascites. Species reactivity: Human. Applications: Flow Cytometry, IP. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-200-7191