Learn More
Wilmad Labglass CLIP RETAINER SZ 1 1-2MM
Supplier:  Wilmad Labglass LG9604100
Encompass
Clip, Retainer, Clip Size: 1, Fits Bore: 1 to 2mm, Used to keep Plug in Place with a Constant Sealing Pressure
Catalog No. 50-802-288