Learn More
Leica Biosystems CLIPPED CORNER SLIDE WH 1440/C
Supplier:  Leica Biosystems 3800080CL
Encompass_Preferred
The Apex Adhesive Slides portfolio consists of microscope slides made of white glass and coated with a proprietary adhesive coating for a positive charge, to promote tissue adhesion and to allow the slide to be more hydrophilic. This hydrophilicity provides a better base for homogeneous covering by aqueous stains and reagents leading to consistent full slide staining. All slides in the Apex portfolio are recommended for demanding procedures such as standard IHC and HIER techniques. Size: 1” x 3” x .04”
Catalog No. NC1721074