Learn More
Medchemexpress LLC Clivatuzumab-MMAE | 98.7% | 10 MG

Supplier: Medchemexpress LLC HY17127010MG
Clivatuzumab-MMAE is an antibody-drug conjugate composed of a MUC1-targeting humanized antibody linked via a valine-citrulline (VC) protease-cleavable linker to the cytotoxic payload monomethyl auristatin E (MMAE). Supplied for research use, the material is shipped on dry ice and stored at -80 °C.
- Antibody-drug conjugate targeting mucin (MUC1).
- Contains a valine-citrulline cleavable linker and MMAE payload.
- Supplied as a liquid and shipped on dry ice.
- Recommended storage at -80 °C; protect from light.
- Includes datasheet, certificate of analysis, and SDS for quality control and handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.