Learn More
Lakeland Industries CLEANMAX CLEAN STERILE Microporous Cleanroom boot. Non-skid sole. 19" upper. Individually packaged in pairs. 50 pr/case with overlay bag. Size 2X
Supplier:  Lakeland Industries CTL903CSP2X
Encompass_Preferred
Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. Let Lakeland clean-manufactured CleanMax apparel be the cornerstone of your contamination control strategy. CleanMax cleanroom garments meet IEST Particle Category I Cleanliness levels. CleanMax sterile products are validated to an SAL 10^-6 with sterile traceability down to the individual garment. 99.999% bacterial and particulate filtration makes CleanMax resistant to microbial, viral and blood borne contaminants, along with most API’s, HD’s, and some cytotoxic drugs. Easy donning fold on each boot. Bound seams provide a superior barrier to prevent breakthrough and strike through. Boots are constructed with a PFA free, fluid-repellant, low-linting, microporous fabric and non-skid sole. Encased elastic and 19" upper. Individually packaged in pairs, 50 pr /case. Size 2XL. Sterile.
Catalog No. NC2957718
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov