Learn More
Lakeland Industries CLEANMAX CLEAN STERILE SMMS Cleanroom coverall. Bound seams, elastic back, wrists, ankles, thumbloops, zipper placket. Individually packaged. 25 per case with overlay bag. Size MD
Supplier:  Lakeland Industries SBC417CSMD
Encompass_Preferred
CleanMax SELECT SMMS Cloth-like fabric Cleanroom Apparel is available in sterile and non-sterile configurations. Clean-Manufactured under strict process and environment controls in Lakeland owned, operated, and certified cleanrooms. CleanMax SELECT cleanroom garments meet IEST Particle Category II Cleanliness levels. CleanMax SELECT sterile products are validated to an SAL 10^-6 w/ sterile traceability down to the individual garment. 96% bacterial and particulate filtration makes CleanMax SELECT resistant to microbial, viral and blood borne contaminants, along w/ most API’s, and HD's. Easy donning fold, zipper plackets, elastic back in the coveralls, thumb loops and always bound seams provide a superior barrier to prevent breakthrough and strike through. Let Lakeland clean-manufactured CleanMax SELECT apparel be the cornerstone of your contamination control strategy. Coveralls are constructed w/ a fluid-repellant, low-linting, four layer breathable SMMS fabric.
Catalog No. NC2945154
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov