Learn More
MSC Rochester Midland RMC Excel NF Non-Foaming Cleaner, Concentrated, 5 gal. Pail, Odorless
Supplier:  MSC 72159676
Encompass
This concentrated cleaner is a water-based alkaline formula with corrosion inhibitors, specifically designed for efficient use in industrial spray washers. It comes in a 5-gallon pail and is formulated for low-foam applications.
  • Water-based alkaline formula
  • Formulated for no foam applications
  • Contains a blend of corrosion inhibitors to protect steel, brass, copper and aluminum
  • Liquid form
  • Odor-free
  • Features sodium metasilicate as the primary chemical
  • Low foam solution
  • 0% VOC content
Catalog No. 50-283-8545