Learn More
AVANOS MEDICAL CLO TEST 25/CS
Supplier:  AVANOS MEDICAL 60480
Encompass
Halyard* Clotest* Rapid Urease Test Is Recognized By Medical Professionals As The "Gold Standard" Among Urease Tests Because Of Its Accuracy, Convenience And Affordability. Clotest Was Developed By Barry Marshall, Md. Who, Along With Robin Warren, M.D. 98% Sensitivity And 97% Specificity Diagnosis Of 75% Of H. Pylori Infections With No False Positives Within 20 Minutes Easy To Read Color Changes Clia Waived
Catalog No. NC9977368
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.