Learn More
Medchemexpress LLC Clobenpropit dihydrobromide | 145231-35-2 | MFCD00467655 | 99.8% | 470.65 g·mol⁻¹ | C14H19Br2ClN4S | 10 MG

Supplier: Medchemexpress LLC HY10119810MG
Clobenpropit dihydrobromide is the dihydrobromide salt of clobenpropit, a histamine receptor ligand used for laboratory research. It acts as a potent histamine H3 receptor antagonist/inverse agonist and a partial H4 agonist, with reported additional binding to 5-HT3 and α2 adrenoceptors. The compound is supplied as a high-purity solid for receptor pharmacology and binding studies.
- High purity (99.76%).
- Molecular formula C14H19Br2ClN4S; molecular weight 470.65 g·mol⁻¹.
- CAS number 145231-35-2.
- Potent H3 antagonist/inverse agonist (pEC50 ~8.07).
- Suitable for receptor pharmacology and binding assays.
- Available in small research quantities (e.g., 10 MG).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.