Learn More
TARGETMOL CHEMICALS INC Clobenpropit dihydrobromide
SDP
Supplier:  TARGETMOL CHEMICALS INC T843910MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Clobenpropit dihydrobromide is a potent antagonist and inverse agonist of histamine H3R (histamine H3LR,pEC50 of 8.07) Purity 99.48%
Catalog No. 50-287-6846