Learn More
Medchemexpress LLC HY-13600 100mg Medchemexpress, Clobetasol propionate CAS:25122-46-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13600100MG
Encompass_Preferred
Medchemexpress, HY-13600 100mg Clobetasol propionate CAS:25122-46-7 Clobetasol propionate is a potent and selective CYP3A5 inhibitor with an IC50 of 0.206 μM. Clobetasol propionate has no inhibiting on CYP3A4 or other major CYPs. Clobetasol propionate is a corticosteroid and has the potential for psoriasis and other dermatoses research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1417