Learn More
Medchemexpress LLC Clobutinol hydrochloride | 1215-83-4 | 99.86% | 292.24 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY148144A100MG
Encompass_Preferred
Clobutinol hydrochloride is a compound with anti-tussive effects. It influences heart rate and blood pressure, making it suitable for cough-related research.
  • Has anti-tussive effects.
  • Influences heart rate and blood pressure.
  • Used in cough related research.
  • Functions as a potassium channel inhibitor.
Catalog No. 50-003-62533