Learn More
Cole-Parmer Digi-Sense Four-Channel, Jumbo-Display Clock/Timer
Supplier:  Cole-Parmer 0864910
Encompass_Preferred
This timer allows you to count up to or down from 100 hours with 1-second resolution. This timer features a 3/4" high display and multiple channels that can be used simultaneously. Each channel has a distinctive alarm that sounds for 1 minute, pressing any button will turn off the alarm. Timer memory recalls previous time set. Features time-of-day in 12- or 24-hour formats.
Catalog No. NC9720376