Learn More
Apexbio Technology LLC Clodronate Liposomes(Synonyms: Clodronate Liposome, Liposomal Clodronate, Clodronate-encapsulated Liposomes), 5mL.
SDP
Supplier:  Apexbio Technology LLC K27215
Encompass_Preferred
Clodronate Liposomes are lipid-based formulations containing encapsulated sodium clodronate designed as macrophage depletion agents for biomedical research Once phagocytosed by macrophages the liposomes release clodronate intracellularly upon lysosomal phospholipase digestion inducing macrophage apoptosis selectively In animal models intravenous administration achieves substantial macrophage depletion in spleen tissue within 24 hours post-injection Researchers employ Clodronate Liposomes to study macrophage functions and related immune-mediated pathologies utilizing various routes such as intravenous intraperitoneal subcutaneous intranasal or direct organ injection
Catalog No. 50-000-61383