Learn More
ENCAPSULA NANAOSCIENCE LLC Clodrosome | 15mL
Supplier:  ENCAPSULA NANAOSCIENCE LLC CLD890915ML
Encompass
Clodrosome is a multilamellar liposome suspension in which clodronate is encapsulated in the aqueous compartments of the liposomes. Encapsome is formulated and prepared identically to Clodrosome except that clodronate is not added to the liposomes. The liposomes are filtered through 2 μm polycarbonate membranes to ensure that the larger particles, which may be toxic to animals, are removed from the suspension. Both are prepared and packaged under sterile conditions. When animals or cells are treated with Clodrosome, phagocytic cells recognize the liposomes as invading foreign particles and proceed to remove the liposomes from the local tissue or serum via phagocytosis. The liposomes then release clodronate into the cytosol, resulting in cell death. Non-encapsulated clodronate cannot cross the cell membrane to initiate cell death.
Catalog No. NC1784900