Learn More
ENCAPSULA NANAOSCIENCE LLC Encapsula Clodrosome, 10 mL
Supplier:  ENCAPSULA NANAOSCIENCE LLC CLD890910ML
Encompass
Clodrosome is a multilamellar liposome suspension in which clodronate is encapsulated in the aqueous compartments of the liposomes.
Catalog No. NC1017326